siRNA Details
| Gene Symbol: D630033A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK052706.1 | - | LIT3440 D630033A02Rik |
192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D630033A02Rik| Gene Symbol: D630033A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK052706.1 | - | LIT3440 D630033A02Rik |
192 | ||