siRNA Details
| Gene Symbol: Gm2044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK144366.1 | - | LIT3443 G630041I22Rik |
567 | ||
| BB748743.1 | - | 97 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB572889.1 | - | 2 | 569 | Chr. 7 - 139957207 - 139958041 [-] on Build GRCm38 | |

Gm2044
Genomic Map