siRNA Details
| Gene Symbol: Gm2044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK144366.1 | - | LIT3443 G630041I22Rik |
756 | ||
| BB748743.1 | - | 286 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2044| Gene Symbol: Gm2044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK144366.1 | - | LIT3443 G630041I22Rik |
756 | ||
| BB748743.1 | - | 286 | ||||