siRNA Details
| Gene Symbol: Linc-md1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK031267.1 | - | 1 | 2798 | ||
| AK132542.1 | - | LIT3444 4831414J02Rik |
1 | 2852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Linc-md1| Gene Symbol: Linc-md1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK031267.1 | - | 1 | 2798 | ||
| AK132542.1 | - | LIT3444 4831414J02Rik |
1 | 2852 | ||