siRNA Details
| Gene Symbol: Zfp264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF365933.1 | - | LIT1344 Zfp264 |
2415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp264| Gene Symbol: Zfp264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF365933.1 | - | LIT1344 Zfp264 |
2415 | ||