siRNA Details
Gene Symbol: LOC102556004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY264286.1 | - | LIT3373 U87HG |
274 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102556004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY264286.1 | - | LIT3373 U87HG |
274 |