siRNA Details
| Gene Symbol: LOC102556004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY264286.1 | - | LIT3373 U87HG |
219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102556004| Gene Symbol: LOC102556004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY264286.1 | - | LIT3373 U87HG |
219 | ||