siRNA Details
| Gene Symbol: LOC287004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110636.1 | 4 | 3507 | |||
| GenBank mRNA | AY035343.1 | - | LIT1742 PINC |
4 | 3506 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC287004| Gene Symbol: LOC287004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110636.1 | 4 | 3507 | |||
| GenBank mRNA | AY035343.1 | - | LIT1742 PINC |
4 | 3506 | |