siRNA Details
| Gene Symbol: LOC287004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110636.1 | 4 | 6677 | |||
| NR_110638.1 | 4 | 2779 | ||||
| NR_110640.1 | 5 | 2110 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DQ099683.1 | - | 5 | 2071 | Chr. 9 - 72706244 - 72723120 [-] on Build Rnor_6.0 | |

LOC287004
Genomic Map