siRNA Details
Gene Symbol: LOC287004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110636.1 | 4 | 6219 | |||
NR_110638.1 | 4 | 2321 | ||||
NR_110640.1 | 5 | 1652 | ||||
NR_110642.1 | 2 | 1054 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC287004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110636.1 | 4 | 6219 | |||
NR_110638.1 | 4 | 2321 | ||||
NR_110640.1 | 5 | 1652 | ||||
NR_110642.1 | 2 | 1054 |