siRNA Details
| Gene Symbol: LOC287004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110637.1 | 1 | 586 | |||
| GenBank mRNA | DQ080211.1 | - | LIT3459 PINC |
1 | 586 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC287004| Gene Symbol: LOC287004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110637.1 | 1 | 586 | |||
| GenBank mRNA | DQ080211.1 | - | LIT3459 PINC |
1 | 586 | |