siRNA Details
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY421730.1 | - | LIT2224 IC-SNURF-SNRPN 3 prime noncoding transcripts |
2, 3 | 266 | |
BC080646.1 | - | 10, 11 | 1393 |
