siRNA Details
| Gene Symbol: SNRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AL832758.1 | - | 11 | 1463 | ||
| AY421730.1 | - | LIT2224 IC-SNURF-SNRPN 3 prime noncoding transcripts |
3 | 345 | ||
| BC080646.1 | - | 11 | 1472 | |||
| BX648788.1 | - | 1 | 23 | |||

SNRPN
Genomic Map