siRNA Details
| Gene Symbol: HNRNPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927544.1 | - | LIT3550 AY927544 |
2 | 348 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HNRNPL| Gene Symbol: HNRNPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927544.1 | - | LIT3550 AY927544 |
2 | 348 | |