siRNA Details
| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927449.1 | - | LIT3477 AY927449 |
4 | 419 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00222| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927449.1 | - | LIT3477 AY927449 |
4 | 419 | |