siRNA Details
| Gene Symbol: SLCO4A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BU741282.1 | - | 1 | 290 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AY927612.3 | - | 1 | 254 | Chr. 20 - 62663047 - 62663718 [+] on Build GRCh38 | |

SLCO4A1-AS1
Genomic Map