siRNA Details
| Gene Symbol: GGTA1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF378121.1 | - | LIT2093 alpha1,3GTP1 |
7 | 710 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

GGTA1P| Gene Symbol: GGTA1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF378121.1 | - | LIT2093 alpha1,3GTP1 |
7 | 710 | |