siRNA Details
| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033376.1 | 4 | 1459 | |||
| GenBank mRNA | AY927448.1 | - | LIT3476 AY927448 |
4 | 1301 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00222| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033376.1 | 4 | 1459 | |||
| GenBank mRNA | AY927448.1 | - | LIT3476 AY927448 |
4 | 1301 | |