siRNA Details
| Gene Symbol: OR52B1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF359415.1 | - | LIT2084 PJCG4 pseudogene |
1 | 59 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52B1P| Gene Symbol: OR52B1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF359415.1 | - | LIT2084 PJCG4 pseudogene |
1 | 59 | |