siRNA Details
| Gene Symbol: OR7E125P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF359416.1 | - | LIT2085 PJCG6 pseudogene |
1 | 151 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR7E125P| Gene Symbol: OR7E125P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF359416.1 | - | LIT2085 PJCG6 pseudogene |
1 | 151 | |