siRNA Details
| Gene Symbol: OR5AK4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_036445.1 | 1 | 685 | |||
| GenBank mRNA | AF309700.1 | - | LIT2083 PJCG3 pseudogene |
1 | 335 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5AK4P| Gene Symbol: OR5AK4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_036445.1 | 1 | 685 | |||
| GenBank mRNA | AF309700.1 | - | LIT2083 PJCG3 pseudogene |
1 | 335 | |