siRNA Details
Gene Symbol: TRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927571.1 | - | LIT3572 AY927571 |
1 | 262 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927571.1 | - | LIT3572 AY927571 |
1 | 262 |