siRNA Details
| Gene Symbol: TRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927571.1 | - | LIT3572 AY927571 |
2 | 706 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAM2| Gene Symbol: TRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927571.1 | - | LIT3572 AY927571 |
2 | 706 | |