siRNA Details
| Gene Symbol: OR7E62P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF359419.1 | - | LIT2088 PJCG9 pseudogene |
11 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR7E62P| Gene Symbol: OR7E62P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF359419.1 | - | LIT2088 PJCG9 pseudogene |
11 | ||