siRNA Details
| Gene Symbol: PCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF103907.1 | - | LIT1552 DD3 |
4 | 1498 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF103908.3 | - | 1 | 3010 | Chr. 9 - 76782136 - 76787569 [+] on Build GRCh38 | |

PCA3
Genomic Map