siRNA Details
| Gene Symbol: HERC2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF041081.1 | - | LIT2191 HERC2P3 |
30, 31 | 5155 | |
| CR749409.1 | - | 23, 24 | 3634 | |||
| Gene Symbol: HERC2P9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036443.1 | 12, 13 | 1817 | |||
| GenBank mRNA | BC047911.1 | - | 12, 13 | 1817 | ||

HERC2P2
Genomic Map