siRNA Details
| Gene Symbol: HERC2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF041081.1 | - | LIT2191 HERC2P3 |
31 | 5294 | |
| CR749409.1 | - | 24 | 3773 | |||
| Gene Symbol: HERC2P9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036443.1 | 13 | 1956 | |||
| GenBank mRNA | AK056876.1 | - | 1 | 1776 | ||
| AL832775.1 | - | 1 | 6319 | |||
| BC047911.1 | - | 13 | 1956 | |||

HERC2P2
Genomic Map