siRNA Details
| Gene Symbol: LINC01276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_120347.1 | 3 | 391 | |||
| GenBank mRNA | AY927638.1 | - | LIT3619 AY927638 |
2 | 235 | |
| DQ141194.1 | - | 3 | 408 | |||

LINC01276
Genomic Map