siRNA Details
| Gene Symbol: LINC01276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927638.1 | - | LIT3619 AY927638 |
1 | 64 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01276| Gene Symbol: LINC01276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927638.1 | - | LIT3619 AY927638 |
1 | 64 | |