siRNA Details
| Gene Symbol: TMEM181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927470.1 | - | 1 | 119 | ||
| AY927471.1 | - | LIT3489 AY927471 |
1 | 119 | ||
| AY927472.1 | - | 1 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM181| Gene Symbol: TMEM181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927470.1 | - | 1 | 119 | ||
| AY927471.1 | - | LIT3489 AY927471 |
1 | 119 | ||
| AY927472.1 | - | 1 | 119 | |||