siRNA Details
| Gene Symbol: SNRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF400496.1 | - | LIT2212 IC-SNURF-SNRPN 3 prime noncoding transcripts |
2, 3 | 416 | |
| AY362865.1 | - | 5, 6 | 457 | |||

SNRPN
Genomic Map