siRNA Details
| Gene Symbol: LINC00184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033927.1 | 2 | 576 | |||
| GenBank mRNA | AJ421679.1 | - | LIT1706 HANC |
1 | 80 | |
| BU950959.1 | - | 2 | 579 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00184| Gene Symbol: LINC00184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033927.1 | 2 | 576 | |||
| GenBank mRNA | AJ421679.1 | - | LIT1706 HANC |
1 | 80 | |
| BU950959.1 | - | 2 | 579 | |||