siRNA Details
Gene Symbol: LINC00184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033927.1 | 2 | 1005 | |||
GenBank mRNA | AJ421679.1 | - | LIT1706 HANC |
1 | 509 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033927.1 | 2 | 1005 | |||
GenBank mRNA | AJ421679.1 | - | LIT1706 HANC |
1 | 509 |