siRNA Details
| Gene Symbol: VN1R108P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF253316.1 | - | LIT2078 PHRET pseudogene |
1 | 1038 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

VN1R108P| Gene Symbol: VN1R108P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF253316.1 | - | LIT2078 PHRET pseudogene |
1 | 1038 | |