siRNA Details
| Gene Symbol: HERC2P7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF071178.2 | - | LIT2070 HERC2P7 pseudogene |
2 | 952 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HERC2P7| Gene Symbol: HERC2P7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF071178.2 | - | LIT2070 HERC2P7 pseudogene |
2 | 952 | |