siRNA Details
| Gene Symbol: LOC101928911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110869.1 | 2 | 142 | |||
| GenBank mRNA | AY927641.1 | - | LIT3622 AY927641 |
2 | 143 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928911| Gene Symbol: LOC101928911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110869.1 | 2 | 142 | |||
| GenBank mRNA | AY927641.1 | - | LIT3622 AY927641 |
2 | 143 | |