siRNA Details
| Gene Symbol: LOC101928911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110869.1 | 4 | 446 | |||
| GenBank mRNA | AY927641.1 | - | LIT3622 AY927641 |
4 | 447 | |
| AY927642.1 | - | 3 | 295 | |||

LOC101928911
Genomic Map