siRNA Details
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927628.1 | - | LIT3614 AY927628 |
2 | 1130 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927628.1 | - | LIT3614 AY927628 |
2 | 1130 |