siRNA Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | XM_005258827.2 | XP_005258884.1 | 2 | 256 | ||
| XM_005258828.2 | XP_005258885.1 | 2 | 277 | |||
| GenBank mRNA | AK097095.1 | - | 1 | 828 | ||
| AY927563.1 | - | 1 | 66 | |||
| AY927628.1 | - | LIT3614 AY927628 |
1 | 66 | ||
| BC000412.2 | - | 1 | 236 | 2961290 | ||
| BC008956.2 | - | 1 | 236 | 2961290 | ||
| GU354322.1 | - | 2 | 69 | |||
| GU354323.1 | - | 2 | 257 | |||

CCDC106
Genomic Map