siRNA Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927628.1 | - | LIT3614 AY927628 |
1203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC106| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927628.1 | - | LIT3614 AY927628 |
1203 | ||