siRNA Details
| Gene Symbol: NPSR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY312370.1 | - | LIT3333 AAA1 |
1, 2 | 87 | |
| BC130477.1 | - | 1, 2 | 87 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPSR1-AS1| Gene Symbol: NPSR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY312370.1 | - | LIT3333 AAA1 |
1, 2 | 87 | |
| BC130477.1 | - | 1, 2 | 87 | |||