siRNA Details
| Gene Symbol: C14orf159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927602.1 | - | LIT3593 AY927602 |
4, 5 | 502 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14orf159| Gene Symbol: C14orf159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927602.1 | - | LIT3593 AY927602 |
4, 5 | 502 | |