siRNA Details
| Gene Symbol: RBMY3AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | U94385.1 | - | LIT2106 RBMIII pseudogene |
3 | 262 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBMY3AP| Gene Symbol: RBMY3AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | U94385.1 | - | LIT2106 RBMIII pseudogene |
3 | 262 | |