siRNA Details
Gene Symbol: ZNF204P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF033199.1 | - | LIT2063 ZNF204 pseudogene |
3 | 1208 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF204P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF033199.1 | - | LIT2063 ZNF204 pseudogene |
3 | 1208 |