siRNA Details
| Gene Symbol: ZNF204P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF033199.1 | - | LIT2063 ZNF204 pseudogene |
2 | 176 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF204P| Gene Symbol: ZNF204P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF033199.1 | - | LIT2063 ZNF204 pseudogene |
2 | 176 | |