siRNA Details
| Gene Symbol: GUSBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | U21555.1 | - | LIT2100 5G4 pseudogene |
6, 7 | 605 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUSBP3| Gene Symbol: GUSBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | U21555.1 | - | LIT2100 5G4 pseudogene |
6, 7 | 605 | |