siRNA Details
| Gene Symbol: STYXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927581.1 | - | LIT3580 AY927581 |
8 | 881 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

STYXL1| Gene Symbol: STYXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927581.1 | - | LIT3580 AY927581 |
8 | 881 | |