siRNA Details
| Gene Symbol: STYXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_001317788.1 | NP_001304717.1 | 5, 6 | 600 | ||
| XM_017012299.1 | XP_016867788.1 | 8, 9 | 998 | |||
| GenBank mRNA | AF169647.1 | - | 2, 3 | 252 | ||
| AK308828.1 | - | 6, 7 | 697 | |||
| AY927581.1 | - | LIT3580 AY927581 |
6, 7 | 677 | ||
| BU859937.1 | - | 5, 6 | 585 | |||

STYXL1
Genomic Map