siRNA Details
| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927446.1 | - | LIT3474 AY927446 |
1 | 88 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00222| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927446.1 | - | LIT3474 AY927446 |
1 | 88 | |