siRNA Details
| Gene Symbol: PRKXP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_073405.1 | 5266 | ||||
| GenBank mRNA | Y10483.1 | - | LIT2111 PRKXP1 |
2 | 456 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKXP1| Gene Symbol: PRKXP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_073405.1 | 5266 | ||||
| GenBank mRNA | Y10483.1 | - | LIT2111 PRKXP1 |
2 | 456 | |